siRNA Details
| Gene Symbol: Mybph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016749.2 | NP_058029.2 | 6, 7 | 1010 | ||
| GenBank mRNA | BC116684.1 | - | 6, 7 | 977 | 40106746 | |
| BC118050.1 | - | 6, 7 | 977 | 40106744 | ||
| CB587371.1 | - | 2, 3 | 257 | |||
| U68267.1 | - | 6, 7 | 1004 | |||

Mybph
Genomic Map