siRNA Details
| Gene Symbol: Mybph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016749.2 | NP_058029.2 | 5, 6 | 857 | ||
| GenBank mRNA | BC116684.1 | - | 5, 6 | 824 | 40106746 | |
| BC118050.1 | - | 5, 6 | 824 | 40106744 | ||
| CB587371.1 | - | 1, 2 | 104 | |||
| U68267.1 | - | 5, 6 | 851 | |||

Mybph
Genomic Map