siRNA Details
| Gene Symbol: Stx7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016797.4 | NP_058077.2 | 2, 3 | 561 | ||
| GenBank mRNA | AF056323.1 | - | 2, 3 | 173 | ||
| AK002867.1 | - | 2, 3 | 230 | |||
| AK013869.1 | - | 2, 3 | 215 | |||
| AK082927.1 | - | 2, 3 | 201 | |||
| AK142742.1 | - | 208 | ||||
| AK158923.1 | - | 2, 3 | 562 | |||
| AK166822.1 | - | 2, 3 | 231 | |||
| AK169822.1 | - | 2, 3 | 234 | |||
| AK210437.1 | - | 1, 2 | 73 | |||
| BC132123.1 | - | 1, 2 | 111 | 40130400 | ||
| BC132125.1 | - | 1, 2 | 111 | 40130402 | ||

Stx7
Genomic Map