siRNA Details
| Gene Symbol: Tuba8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017379.1 | NP_059075.1 | 4 | 865 | ||
| GenBank mRNA | AJ245923.1 | - | 4 | 865 | ||
| AK009439.1 | - | 2 | 450 | |||
| AK009651.1 | - | 2 | 451 | |||
| AK032157.1 | - | 838 | ||||
| BC017631.1 | - | 4 | 844 | 4507364 | ||
| GU591980.1 | - | 817 | ||||

Tuba8
Genomic Map