siRNA Details
| Gene Symbol: Gpatch11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181649.6 | NP_857632.4 | 4, 5 | 597 | ||
| GenBank mRNA | AK009084.1 | - | 4, 5 | 484 | ||
| AK148331.1 | - | 4, 5 | 599 | |||
| BC047998.1 | - | 4, 5 | 511 | 6511097 | ||
| BC092522.1 | - | 4, 5 | 524 | 30648550 | ||
| BC138310.1 | - | 4, 5 | 492 | 8861330 | ||
| BY741452.1 | - | 4, 5 | 598 | |||

Gpatch11
Genomic Map