siRNA Details
Gene Symbol: Gpatch11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181649.6 | NP_857632.4 | 6 | 650 | ||
GenBank mRNA | AK009084.1 | - | 6 | 537 | ||
BC092522.1 | - | 6 | 577 | 30648550 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gpatch11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181649.6 | NP_857632.4 | 6 | 650 | ||
GenBank mRNA | AK009084.1 | - | 6 | 537 | ||
BC092522.1 | - | 6 | 577 | 30648550 |