siRNA Details
| Gene Symbol: Rfx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017395.2 | NP_059091.2 | 1 | 31 | ||
| GenBank mRNA | AB221679.1 | - | 1 | 12 | ||
| AF209854.1 | - | 1 | 12 | |||
| AK088686.1 | - | 2 | 83 | |||
| BB854843.1 | - | 2 | 124 | |||
| BC051965.1 | - | 2 | 78 | 5702640 | ||
| BC057336.1 | - | 2 | 57 | |||
| BC075717.1 | - | 3 | 140 | |||
| BC118948.2 | - | 1 | 12 | 40092396 | ||
| BC118949.2 | - | 1 | 12 | 40092397 | ||
| BY020562.1 | - | 2 | 103 | |||
| BY230827.1 | - | 2 | 63 | |||

Rfx5
Genomic Map