siRNA Details
| Gene Symbol: Rfx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017395.2 | NP_059091.2 | 7 | 742 | ||
| GenBank mRNA | AB221679.1 | - | 7 | 723 | ||
| AF209854.1 | - | 7 | 723 | |||
| AK088686.1 | - | 7 | 908 | |||
| BC051965.1 | - | 8 | 779 | 5702640 | ||
| BC057336.1 | - | 8 | 768 | |||
| BC075717.1 | - | 9 | 851 | |||
| BC118948.2 | - | 7 | 723 | 40092396 | ||
| BC118949.2 | - | 7 | 723 | 40092397 | ||

Rfx5
Genomic Map