siRNA Details
| Gene Symbol: Icos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017480.2 | NP_059508.2 | 3 | 495 | ||
| GenBank mRNA | AB023132.1 | - | 3 | 479 | ||
| AF216748.1 | - | 3 | 450 | |||
| AF257230.1 | - | 3 | 480 | |||
| AJ250559.1 | - | 3 | 495 | |||
| AK030827.1 | - | 3 | 488 | |||
| BC034852.1 | - | 3 | 482 | 1364228 | ||

Icos
Genomic Map