siRNA Details
| Gene Symbol: Rfk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019437.3 | NP_062310.1 | 2 | 368 | ||
| GenBank mRNA | AK002806.1 | - | 2 | 329 | ||
| AK008352.1 | - | 2 | 298 | |||
| AK010607.1 | - | 2 | 297 | |||
| BC033521.1 | - | 2 | 270 | 4527980 | ||
| BC051021.1 | - | 2 | 279 | 6315425 | ||

Rfk
Genomic Map