siRNA Details
| Gene Symbol: Dll4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019454.3 | NP_062327.2 | 9 | 1881 | ||
| GenBank mRNA | AB043893.1 | - | 9 | 1524 | ||
| AF253469.1 | - | 9 | 1836 | |||
| AF273454.1 | - | 9 | 1902 | |||
| AK004739.1 | - | 9 | 1849 | |||
| AK031699.1 | - | 9 | 1851 | |||
| AK052322.1 | - | 9 | 1883 | |||
| AK165517.1 | - | 9 | 1849 | |||
| AK171717.1 | - | 9 | 1848 | |||
| BC042497.1 | - | 9 | 1836 | 4017786 | ||
| BC049130.1 | - | 9 | 1841 | 6825525 | ||

Dll4
Genomic Map