siRNA Details
| Gene Symbol: Eef1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018796.3 | NP_061266.2 | 3 | 679 | ||
| GenBank mRNA | AF029844.2 | - | 3 | 345 | ||
| AK012756.1 | - | 3 | 382 | |||
| AK027911.1 | - | 3 | 382 | |||
| AK168191.1 | - | 3 | 381 | |||
| BC003899.1 | - | 3 | 375 | 3600850 | ||
| BC023139.1 | - | 3 | 391 | 3963930 | ||
| BC039635.1 | - | 3 | 394 | |||

Eef1b2
Genomic Map