siRNA Details
| Gene Symbol: Cts8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019541.3 | NP_062414.3 | 7 | 953 | ||
| GenBank mRNA | AF250840.1 | - | 7 | 965 | ||
| AK049474.1 | - | 7 | 954 | |||
| AY014780.1 | - | 7 | 952 | |||
| BC068241.1 | - | 7 | 943 | 30019120 | ||

Cts8
Genomic Map