siRNA Details
| Gene Symbol: Cts8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019541.3 | NP_062414.3 | 7, 8 | 981 | ||
| GenBank mRNA | AF250840.1 | - | 7, 8 | 993 | ||
| AK049474.1 | - | 7, 8 | 982 | |||
| AY014780.1 | - | 7, 8 | 980 | |||
| BC068241.1 | - | 7, 8 | 971 | 30019120 | ||

Cts8
Genomic Map