siRNA Details
| Gene Symbol: Nagk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164187.1 | NP_001157659.1 | 5 | 574 | ||
| NM_019542.2 | NP_062415.1 | 5 | 583 | |||
| GenBank mRNA | AJ242909.1 | - | 5 | 595 | ||
| AJ242911.1 | - | 5 | 535 | |||
| AK007240.1 | - | 5 | 578 | |||
| AK155292.1 | - | 5 | 475 | |||
| BC004689.1 | - | 5 | 461 | 3501470 | ||

Nagk
Genomic Map