siRNA Details
| Gene Symbol: Scamp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019575.4 | NP_062521.1 | 7 | 727 | ||
| GenBank mRNA | AF224721.1 | - | 7 | 702 | ||
| AF295102.1 | - | 7 | 720 | |||
| AK019112.1 | - | 7 | 731 | |||
| AK078226.1 | - | 7 | 719 | |||
| AK154094.1 | - | 7 | 718 | |||
| AK160755.1 | - | 7 | 718 | |||
| AK168554.1 | - | 7 | 713 | |||
| BC018215.1 | - | 7 | 696 | 3965223 | ||
| BC054104.2 | - | 7 | 722 | 30021104 | ||

Scamp4
Genomic Map