siRNA Details
| Gene Symbol: Pex14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019781.2 | NP_062755.1 | 6, 7 | 496 | ||
| GenBank mRNA | AF097512.1 | - | 6, 7 | 496 | ||
| AK148779.1 | - | 5, 6 | 3423 | |||
| BC028952.1 | - | 6, 7 | 493 | 5368312 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pex14| Gene Symbol: Pex14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019781.2 | NP_062755.1 | 6, 7 | 496 | ||
| GenBank mRNA | AF097512.1 | - | 6, 7 | 496 | ||
| AK148779.1 | - | 5, 6 | 3423 | |||
| BC028952.1 | - | 6, 7 | 493 | 5368312 | ||