siRNA Details
| Gene Symbol: Rab9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019773.2 | NP_062747.1 | 3 | 825 | ||
| GenBank mRNA | AB027290.1 | - | 3 | 654 | ||
| AB232602.1 | - | 1 | 439 | |||
| AK010710.1 | - | 3 | 604 | |||
| AK017301.1 | - | 3 | 619 | |||
| AK032133.1 | - | 3 | 590 | |||
| BC008160.1 | - | 3 | 624 | 3964134 | ||

Rab9
Genomic Map