siRNA Details
| Gene Symbol: Trip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019797.4 | NP_062771.2 | 15 | 2606 | ||
| GenBank mRNA | AF197574.1 | - | 12 | 1665 | ||
| AF539614.1 | - | 15 | 2489 | |||
| AK146298.1 | - | 12 | 1673 | |||
| BC021316.1 | - | 12 | 1678 | 5027899 | ||

Trip4
Genomic Map