siRNA Details
| Gene Symbol: Vapb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019806.5 | NP_062780.2 | 5, 6 | 696 | ||
| GenBank mRNA | AF115504.1 | - | 5, 6 | 574 | ||
| AK050632.1 | - | 5, 6 | 645 | |||
| AK081908.1 | - | 5, 6 | 625 | |||
| AK160925.1 | - | 5, 6 | 539 | |||
| BC048231.1 | - | 5, 6 | 696 | 6313869 | ||
| BC057123.1 | - | 5, 6 | 685 | 6411688 | ||

Vapb
Genomic Map