siRNA Details
| Gene Symbol: Nme4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019731.1 | NP_062705.1 | 4 | 389 | ||
| GenBank mRNA | AF153451.1 | - | 4 | 389 | ||
| AF288690.1 | - | 4 | 402 | |||
| AK011573.1 | - | 2 | 699 | |||
| AK012805.1 | - | 4 | 425 | |||
| AK017724.1 | - | 4 | 405 | |||
| BC027277.1 | - | 4 | 407 | 3591383 | ||

Nme4
Genomic Map