siRNA Details
| Gene Symbol: Sec11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019951.1 | NP_064335.1 | 5 | 556 | ||
| GenBank mRNA | AB025405.1 | - | 5 | 556 | ||
| AK007449.1 | - | 5 | 554 | |||
| BC010484.1 | - | 5 | 548 | 3483051 | ||
| BC057885.1 | - | 5 | 748 | |||

Sec11a
Genomic Map