siRNA Details
| Gene Symbol: Habp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019986.3 | NP_064370.2 | 4, 5 | 850 | ||
| GenBank mRNA | AF227684.1 | - | 4, 5 | 793 | ||
| AK016800.1 | - | 4, 5 | 893 | |||
| AK144206.1 | - | 4, 5 | 834 | |||
| AK146525.1 | - | 4, 5 | 823 | |||
| BC082806.1 | - | 4, 5 | 852 | |||
| BC119460.2 | - | 3, 4 | 347 | |||
| BC148304.2 | - | 3, 4 | 354 | |||

Habp4
Genomic Map