siRNA Details
| Gene Symbol: Mocs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020042.2 | NP_064426.2 | 11 | 1936 | ||
| NM_028464.1 | NP_082740.1 | 11 | 1951 | |||
| GenBank mRNA | AK157231.1 | - | 11 | 1892 | ||
| BC027444.1 | - | 3 | 862 | |||
| BC034066.1 | - | 7 | 1300 | |||
| BC049914.1 | - | 9 | 1579 | |||
| BC147411.1 | - | 2 | 780 | |||
| BC147412.1 | - | 2 | 765 | |||
| BC172067.1 | - | 2 | 765 | |||

Mocs1
Genomic Map