siRNA Details
| Gene Symbol: Tmem9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020050.1 | NP_064434.1 | 3 | 332 | ||
| GenBank mRNA | AK007430.1 | - | 3 | 300 | ||
| AK009147.1 | - | 3 | 267 | |||
| AK049882.1 | - | 3 | 419 | |||
| AK077253.1 | - | 3 | 288 | |||
| AK147127.1 | - | 3 | 348 | |||
| AK201035.1 | - | 2 | 146 | |||
| AK211319.1 | - | 2 | 146 | |||
| BC002208.1 | - | 3 | 316 | 3490055 | ||

Tmem9b
Genomic Map