siRNA Details
| Gene Symbol: Tmem9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020050.1 | NP_064434.1 | 2 | 249 | ||
| GenBank mRNA | AK007430.1 | - | 2 | 217 | ||
| AK009147.1 | - | 2 | 184 | |||
| AK049882.1 | - | 2 | 336 | |||
| AK077253.1 | - | 2 | 205 | |||
| AK201035.1 | - | 1 | 63 | |||
| AK211319.1 | - | 1 | 63 | |||
| BC002208.1 | - | 2 | 233 | 3490055 | ||

Tmem9b
Genomic Map