siRNA Details
| Gene Symbol: Gmeb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122992.1 | NP_001116464.1 | 5 | 433 | ||
| NM_020273.2 | NP_064669.2 | 4 | 370 | |||
| GenBank mRNA | AF210433.1 | - | 3 | 331 | ||
| AK134349.1 | - | 4 | 371 | |||
| BC046983.1 | - | 5 | 404 | 6477130 | ||
| BU701415.1 | - | 3 | 217 | |||
| BY732360.1 | - | 5 | 408 | |||
| DV660282.1 | - | 3 | 288 | |||

Gmeb1
Genomic Map