siRNA Details
| Gene Symbol: Ctsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020284.1 | NP_064680.1 | 2 | 133 | ||
| GenBank mRNA | AF245399.1 | - | 2 | 133 | ||
| AK005429.1 | - | 2 | 153 | |||
| AK014432.1 | - | 2 | 149 | |||
| AY014778.1 | - | 2 | 92 | |||
| AY943668.1 | - | 2 | 202 | |||
| BC100338.1 | - | 2 | 128 | 30712889 | ||

Ctsr
Genomic Map