siRNA Details
| Gene Symbol: Ctsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020284.1 | NP_064680.1 | 4, 5 | 479 | ||
| GenBank mRNA | AF245399.1 | - | 4, 5 | 479 | ||
| AK005429.1 | - | 4, 5 | 499 | |||
| AK014432.1 | - | 4, 5 | 495 | |||
| AY014778.1 | - | 4, 5 | 438 | |||
| AY943668.1 | - | 4, 5 | 551 | |||
| BC100338.1 | - | 4, 5 | 474 | 30712889 | ||

Ctsr
Genomic Map