siRNA Details
| Gene Symbol: Tob2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020507.3 | NP_065253.1 | 2 | 529 | ||
| GenBank mRNA | AK049766.1 | - | 2 | 476 | ||
| AK122537.1 | - | 1 | 1734 | |||
| AK143542.1 | - | 2 | 506 | |||
| BC011163.1 | - | 1 | 411 | 4194136 | ||
| BC021869.1 | - | 2 | 463 | |||
| BC094031.1 | - | 2 | 460 | 4945656 | ||

Tob2
Genomic Map