siRNA Details
| Gene Symbol: Retnla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020509.3 | NP_065255.2 | 2 | 189 | ||
| GenBank mRNA | AF205951.1 | - | 1 | 122 | ||
| AF290872.1 | - | 2 | 227 | |||
| AF316397.2 | - | 2 | 131 | |||
| AF323082.1 | - | 2 | 199 | |||
| AK007553.1 | - | 2 | 134 | |||
| BC029248.1 | - | 1 | 129 | 4189285 | ||

Retnla
Genomic Map