siRNA Details
| Gene Symbol: Park7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020569.3 | NP_065594.2 | 4 | 388 | ||
| GenBank mRNA | AB015652.1 | - | 4 | 335 | ||
| AK146368.1 | - | 4 | 300 | |||
| AK153948.1 | - | 4 | 345 | |||
| AK168341.1 | - | 4 | 318 | |||
| BC002187.1 | - | 4 | 294 | 3487763 | ||
| CA456587.1 | - | 4 | 426 | |||
| CD776363.1 | - | 4 | 529 | |||

Park7
Genomic Map