siRNA Details
| Gene Symbol: Gsdma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021347.4 | NP_067322.1 | 9 | 905 | ||
| GenBank mRNA | AB033595.1 | - | 9 | 894 | ||
| AK028645.1 | - | 9 | 906 | |||
| AK028698.1 | - | 9 | 906 | |||
| AK029062.1 | - | 9 | 889 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsdma| Gene Symbol: Gsdma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021347.4 | NP_067322.1 | 9 | 905 | ||
| GenBank mRNA | AB033595.1 | - | 9 | 894 | ||
| AK028645.1 | - | 9 | 906 | |||
| AK028698.1 | - | 9 | 906 | |||
| AK029062.1 | - | 9 | 889 | |||