siRNA Details
Gene Symbol: Sertad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038625.1 | NP_001033714.1 | 2 | 937 | ||
NM_021372.2 | NP_067347.2 | 2 | 1148 | |||
GenBank mRNA | AB041541.1 | - | 2 | 1067 | ||
AF366403.1 | - | 1 | 252 | |||
AK076787.1 | - | 2 | 841 | |||
AK129059.1 | - | 1 | 97 | |||
AK132846.1 | - | 2 | 917 | |||
AK132849.1 | - | 2 | 756 | |||
AK142018.1 | - | 2 | 917 | |||
AK146621.1 | - | 2 | 938 | |||
AK146897.1 | - | 2 | 939 | |||
AK147674.1 | - | 3 | 1324 | |||
BC008257.1 | - | 1 | 94 | |||
BC014726.1 | - | 2 | 980 | 4240282 |
