siRNA Details
| Gene Symbol: Sectm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026907.3 | NP_081183.3 | 4, 5 | 631 | ||
| GenBank mRNA | AK007321.1 | - | 3, 4 | 493 | ||
| AK029082.1 | - | 4, 5 | 632 | |||
| AK143807.1 | - | 4, 5 | 615 | |||
| BC020159.1 | - | 3, 4 | 582 | 4016267 | ||

Sectm1b
Genomic Map