siRNA Details
| Gene Symbol: Rab37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163753.1 | NP_001157225.1 | 4, 5 | 723 | ||
| NM_021411.4 | NP_067386.3 | 4, 5 | 335 | |||
| GenBank mRNA | AB232633.1 | - | 4, 5 | 314 | ||
| AF233582.1 | - | 4, 5 | 314 | |||
| AK039716.1 | - | 4, 5 | 329 | |||
| AK040216.1 | - | 4, 5 | 335 | |||
| AK045995.1 | - | 4, 5 | 326 | |||
| AK046260.1 | - | 4, 5 | 724 | |||
| AK050483.1 | - | 4, 5 | 329 | |||
| BC050757.1 | - | 4, 5 | 335 | |||
| BC128020.1 | - | 4, 5 | 333 | 40136290 | ||

Rab37
Genomic Map