siRNA Details
| Gene Symbol: Stk25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021537.3 | NP_067512.3 | 7 | 993 | ||
| GenBank mRNA | AF004934.1 | - | 6 | 831 | ||
| AK148041.1 | - | 7 | 967 | |||
| AK152917.1 | - | 7 | 936 | |||
| BC029875.1 | - | 6 | 1711 | |||
| BC052913.1 | - | 7 | 928 | 30057101 | ||
| BC071218.1 | - | 7 | 988 | 4012695 | ||

Stk25
Genomic Map