siRNA Details
| Gene Symbol: Mettl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021554.2 | NP_067529.2 | 2 | 580 | ||
| GenBank mRNA | AB041664.1 | - | 2 | 358 | ||
| AJ278576.1 | - | 2 | 580 | |||
| AK075586.1 | - | 2 | 343 | |||
| AK201397.1 | - | 2 | 68 | |||
| BC023188.1 | - | 1 | 21 | |||
| BC068124.1 | - | 2 | 374 | 5716972 | ||

Mettl9
Genomic Map