siRNA Details
| Gene Symbol: Cenpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021790.2 | NP_068562.1 | 2, 3 | 414 | ||
| NM_181061.5 | NP_851406.1 | 2, 3 | 537 | |||
| GenBank mRNA | AB043687.1 | - | 2, 3 | 414 | ||
| AK082939.1 | - | 2, 3 | 194 | |||
| AK160776.1 | - | 2, 3 | 172 | |||
| BC028655.1 | - | 2, 3 | 537 | |||
| BC092351.1 | - | 2, 3 | 176 | |||

Cenpk
Genomic Map