siRNA Details
| Gene Symbol: Aqp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271843.1 | NP_001258772.1 | 5 | 838 | ||
| NM_022026.3 | NP_071309.1 | 5 | 838 | |||
| GenBank mRNA | AB037180.1 | - | 5 | 635 | ||
| AK050042.1 | - | 5 | 838 | |||
| AK154151.1 | - | 5 | 842 | |||
| BC024105.1 | - | 6 | 959 | 5096952 | ||
| CT010174.1 | - | 5 | 632 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK050325.1 | - | 1894 | Chr. - - [-] on Build | ||

Aqp9
Genomic Map