siRNA Details
Gene Symbol: Inpp5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290437.1 | NP_001277366.1 | 2 | 829 | ||
NM_033134.3 | NP_149125.1 | 1 | 1113 | |||
GenBank mRNA | AF226683.1 | - | 1 | 1075 | ||
AK047245.1 | - | 1 | 1098 | |||
AK080075.1 | - | 1 | 1095 | |||
AK085439.1 | - | 2 | 832 | |||
AK154097.1 | - | 1 | 1114 | |||
AK158135.1 | - | 1 | 1098 | |||
AK170174.1 | - | 1 | 1046 | |||
AK170786.1 | - | 1 | 1070 | |||
AK170812.1 | - | 1 | 1084 | |||
BC052717.1 | - | 1 | 1082 | 6837339 | ||
BC057566.1 | - | 1 | 1078 | |||
BC080295.1 | - | 1 | 1078 | 30354397 |
