siRNA Details
| Gene Symbol: Irak1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022986.4 | NP_075362.3 | 2, 3 | 601 | ||
| GenBank mRNA | AF093135.1 | - | 2, 3 | 384 | ||
| AJ242722.1 | - | 2, 3 | 226 | |||
| AK013036.2 | - | 2, 3 | 406 | |||
| AK020170.1 | - | 2, 3 | 604 | |||
| AK020448.1 | - | 2, 3 | 376 | |||
| AK045436.1 | - | 2, 3 | 397 | |||
| AK076594.1 | - | 2, 3 | 409 | |||
| BC046301.1 | - | 2, 3 | 395 | |||
| BC061099.1 | - | 2, 3 | 388 | 6742251 | ||

Irak1bp1
Genomic Map