siRNA Details
| Gene Symbol: Zfp524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025324.2 | NP_079600.1 | 2 | 953 | ||
| GenBank mRNA | AK002594.1 | - | 2 | 949 | ||
| AK009053.1 | - | 2 | 949 | |||
| AK011620.1 | - | 2 | 954 | |||
| AK154328.1 | - | 2 | 960 | |||
| AK217603.1 | - | 1 | 14 | |||
| BC019995.1 | - | 2 | 966 | 4458724 | ||

Zfp524
Genomic Map