siRNA Details
| Gene Symbol: Tmem176a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098271.1 | NP_001091741.1 | 5 | 442 | ||
| NM_025326.4 | NP_079602.4 | 5 | 494 | |||
| GenBank mRNA | AB063313.1 | - | 4 | 384 | ||
| AK002546.1 | - | 5 | 470 | |||
| AK079378.1 | - | 456 | ||||
| AK150576.1 | - | 5 | 459 | |||
| AK162991.1 | - | 3 | 1702 | |||
| AK165660.1 | - | 5 | 468 | |||
| BC010831.1 | - | 5 | 438 | 4221626 | ||
| CT010190.1 | - | 4 | 384 | |||

Tmem176a
Genomic Map