siRNA Details
| Gene Symbol: Emc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175101.3 | NP_780310.1 | 8 | 928 | ||
| GenBank mRNA | AK020058.1 | - | 8 | 899 | ||
| AK075595.1 | - | 8 | 929 | |||
| AK167819.1 | - | 8 | 777 | |||
| BC004641.1 | - | 8 | 924 | 3157147 | ||

Emc3
Genomic Map