siRNA Details
Gene Symbol: Grina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023168.3 | NP_075657.1 | 4 | 628 | ||
GenBank mRNA | AF182040.1 | - | 4 | 611 | ||
AK003920.1 | - | 4 | 636 | |||
AK189399.1 | - | 3 | 252 | |||
BC010802.1 | - | 4 | 661 | 4236429 | ||
BC019157.1 | - | 4 | 649 | |||
BC037667.1 | - | 4 | 628 | 5376054 |
