siRNA Details
| Gene Symbol: Grina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023168.3 | NP_075657.1 | 3 | 458 | ||
| GenBank mRNA | AF182040.1 | - | 3 | 441 | ||
| AK003920.1 | - | 3 | 467 | |||
| AK179160.1 | - | 2 | 82 | |||
| AK189399.1 | - | 2 | 82 | |||
| AK200011.1 | - | 2 | 109 | |||
| AK212726.1 | - | 2 | 125 | |||
| BC010802.1 | - | 3 | 491 | 4236429 | ||
| BC019157.1 | - | 3 | 479 | |||
| BC037667.1 | - | 3 | 458 | 5376054 | ||

Grina
Genomic Map