siRNA Details
Gene Symbol: Ssr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025448.3 | NP_079724.1 | 1 | 292 | ||
GenBank mRNA | AK005340.1 | - | 2 | 163 | ||
AK010565.1 | - | 2 | 130 | |||
AK020145.1 | - | 2 | 113 | |||
AK050505.1 | - | 1 | 293 | |||
AK084530.1 | - | 2 | 130 | |||
AK185121.1 | - | 1 | 70 | |||
BC010214.1 | - | 1 | 276 | 3497894 |
