siRNA Details
| Gene Symbol: Ssr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025448.3 | NP_079724.1 | 1, 2 | 371 | ||
| GenBank mRNA | AK005340.1 | - | 2, 3 | 242 | ||
| AK010565.1 | - | 2, 3 | 209 | |||
| AK012281.1 | - | 1, 2 | 199 | |||
| AK020145.1 | - | 2, 3 | 192 | |||
| AK050505.1 | - | 1, 2 | 372 | |||
| AK084530.1 | - | 2, 3 | 209 | |||
| BC010214.1 | - | 1, 2 | 355 | 3497894 | ||

Ssr2
Genomic Map