siRNA Details
| Gene Symbol: Nicn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025449.3 | NP_079725.1 | 3 | 467 | ||
| GenBank mRNA | AJ299741.1 | - | 3 | 459 | ||
| AK002789.1 | - | 3 | 473 | |||
| AK013602.1 | - | 3 | 495 | |||
| AK075732.1 | - | 3 | 475 | |||
| AK146890.1 | - | 3 | 477 | |||
| AK169135.1 | - | 3 | 477 | |||
| BC024050.1 | - | 3 | 467 | 5133763 | ||

Nicn1
Genomic Map